4-Hydroxy-3,5-diiodobenzonitrile ≥97%

Supplier: Matrix Scientific

084484-1G 084484-10G
102842-536EA 101.54 USD
102842-536 102842-538
4-Hydroxy-3,5-diiodobenzonitrile ≥97%
4-Hydroxy-3,5-diiodobenzonitrile

Hazard: Irritant

Formula: C₇H₃I₂NO
MDL Number: MFCD00016422
CAS Number: 1689-83-4
UN: 3439
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR