(3,4-Dimethoxyphenylthio)acetic acid ≥97%

Supplier: Matrix Scientific

084481-1G 084481-10G 084481-50G
102842-526EA 113.85 USD
102842-526 102842-528 102842-530
(3,4-Dimethoxyphenylthio)acetic acid ≥97%
(3,4-Dimethoxyphenylthio)acetic acid

Hazard: Irritant

Formula: C₁₀H₁₂O₄S
MDL Number: MFCD00052112
CAS Number: 95735-63-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR