N-(5-Chloro-2-methylphenyl)acetamide ≥97%

Supplier: Matrix Scientific

084477-1G 084477-10G
102842-516EA 255.38 USD
102842-516 102842-518
N-(5-Chloro-2-methylphenyl)acetamide ≥97%
N-(5-Chloro-2-methylphenyl)acetamide

Hazard: Irritant

Formula: C₉H₁₀ClNO
MDL Number: MFCD00018743
CAS Number: 5900-55-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR