1-Isopropenyl-4-(trifluoromethyl)benzene ≥97%

Supplier: Matrix Scientific

084476-1G 084476-10G
102842-512EA 200 USD
102842-512 102842-514
1-Isopropenyl-4-(trifluoromethyl)benzene ≥97%
1-Isopropenyl-4-(trifluoromethyl)benzene

Hazard: Irritant

Formula: C₁₀H₉F₃
MDL Number: MFCD08457470
CAS Number: 55186-75-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR