Thiobenzamide ≥97%

Supplier: Matrix Scientific

Synonyms: Benzothioamide

084473-1G 084473-10G 084473-50G
102842-500EA 1473.85 USD
102842-500 102842-502 102842-504
Thiobenzamide ≥97%
Thiobenzamide

TCA listed: T
Hazard: Toxic

Formula: C₇H₇NS
MW: 137.21 g/mol
Melting Pt: 114-118
Flash Pt: 120°C
MDL Number: MFCD00008060
CAS Number: 2227-79-4
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR