1-Phenyl-3-(trimethylsilyl)-2-propyn-1-ol ≥97%

Supplier: Matrix Scientific

Synonyms: α-[2-(Trimethylsilyl)ethynyl]benzenemethanol

084471-1G 084471-10G
102842-496EA 226.15 USD
102842-496 102842-498
1-Phenyl-3-(trimethylsilyl)-2-propyn-1-ol ≥97%
1-Phenyl-3-(trimethylsilyl)-2-propyn-1-ol

Hazard: Irritant

Formula: C₁₂H₁₆OSi
MDL Number: MFCD05864336
CAS Number: 89530-34-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR