Allyl phenyl sulfone ≥97%
Supplier: Matrix Scientific
Synonyms:
Phenyl 2-propenyl sulfone, Allylsulfonylbenzene, Allyl Phenyl sulphone, (Allylsulphonyl)benzene, (2-Propen-1-ylsulfonyl)benzene, (2-Propenylsulfonyl)benzene
084470-1G
084470-10G
084470-50G
102842-490EA
44.62
USD
102842-490
102842-492
102842-494
Allyl phenyl sulfone ≥97%
Allyl phenyl sulphone
Hazard: Irritant
Formula:
C₉H₁₀O₂S MW: 182.24 g/mol |
MDL Number:
MFCD00014739 CAS Number: 16212-05-8 |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...