4'-(1-Piperazinyl)acetophenone ≥97%

Supplier: Matrix Scientific

084467-1G 084467-10G 084467-50G
102842-480EA 44.62 USD
102842-480 102842-482 102842-484
4'-(1-Piperazinyl)acetophenone ≥97%
4'-(1-Piperazinyl)acetophenone

Hazard: Irritant

Formula: C₁₂H₁₆N₂O
MDL Number: MFCD00005962
CAS Number: 51639-48-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR