5-Methyl-2-(trifluoromethyl)-3-furansulfonyl chloride ≥97%

Supplier: Matrix Scientific

084440-1G 084440-10G
102842-418EA 353.85 USD
102842-418 102842-420
5-Methyl-2-(trifluoromethyl)-3-furansulfonyl chloride ≥97%
5-Methyl-2-(trifluoromethyl)-3-furansulfonyl chloride

Hazard: Irritant

Formula: C₆H₄ClF₃O₃S
MDL Number: MFCD02677699
CAS Number: 306935-02-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR