2-Bromo-4,5-difluorobenzenesulfonyl chloride ≥95%
Supplier: Matrix Scientific
084428-1G
084428-10G
102842-384EA
213.85
USD
102842-384
102842-386
2-Bromo-4,5-difluorobenzenesulfonyl chloride ≥95%
2-Bromo-4,5-difluorobenzenesulfonyl chloride
Hazard: Irritant
Formula:
C₆H₂BrClF₂O₂S |
MDL Number:
MFCD04035650 CAS Number: 874784-11-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...