3,5-Dichloro-2-hydroxybenzenesulfonyl chloride ≥97%

Supplier: Matrix Scientific

084427-1G 084427-10G
102842-380EA 100 USD
102842-380 102842-382
3,5-Dichloro-2-hydroxybenzenesulfonyl chloride ≥97%
3,5-Dichloro-2-hydroxybenzenesulphonyl chloride

Hazard: Corrosive/Moisture Sensitive

Formula: C₆H₃Cl₃O₃S
Melting Pt: 78…80 °C
MDL Number: MFCD00007432
CAS Number: 23378-88-3
UN: 3261
ADR: 8,I

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR