(3-(Trifluoromethyl)phenyl)methanesulfonyl chloride ≥97%

Supplier: Matrix Scientific

Synonyms: 3-(Trifluoromethyl)benzylsulphonyl chloride

084421-1G 084421-10G
102842-362EA 241.54 USD
102842-362 102842-364
(3-(Trifluoromethyl)phenyl)methanesulfonyl chloride ≥97%
(3-(Trifluoromethyl)phenyl)methanesulphonyl chloride

Hazard: Irritant

Formula: C₈H₆ClF₃O₂S
MDL Number: MFCD01631926
CAS Number: 127162-96-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR