[4-(4-Chlorophenyl)phenyl]sulfonyl chloride ≥97%

Supplier: Matrix Scientific

Synonyms: 4'-Chloro-[1,1'-biphenyl]-4-sulphonyl chloride, 4'-Chlorobiphenyl-4-sulphonyl chloride

084407-1G 084407-10G
102842-330EA 226.15 USD
102842-330 102842-332
[4-(4-Chlorophenyl)phenyl]sulfonyl chloride ≥97%
[4-(4-Chlorophenyl)phenyl]sulfonyl chloride

Hazard: Irritant

Formula: C₁₂H₈Cl₂O₂S
MDL Number: MFCD01631918
CAS Number: 20443-74-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR