3',4'-Dichlorobiphenyl-4-sulfonyl chloride ≥97%

Supplier: Matrix Scientific

Synonyms: [4-(3,4-Dichlorophenyl)phenyl]sulphonyl chloride

084406-1G 084406-10G
102842-326EA 383.08 USD
102842-326 102842-328
3',4'-Dichlorobiphenyl-4-sulfonyl chloride ≥97%
3',4'-Dichlorobiphenyl-4-sulfonyl chloride

Hazard: Irritant

Formula: C₁₂H₇Cl₃O₂S
MDL Number: MFCD01631920
CAS Number: 501697-80-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR