Biphenyl-4-sulfonyl chloride ≥95%

Supplier: Matrix Scientific

Synonyms: Biphenyl-4-sulphonyl Chloride

084402-10G 084402-50G
102842-314EA 310.77 USD
102842-314 102842-316
Biphenyl-4-sulfonyl chloride ≥95%
Biphenyl-4-sulphonyl chloride

Hazard: Irritant

Formula: C₁₂H₉ClO₂S
MW: 252.72 g/mol
MDL Number: MFCD00078658
CAS Number: 1623-93-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR