3-Chloro-4-(trifluoromethoxy)benzenesulfonyl chloride ≥95%

Supplier: Matrix Scientific

084401-1G 084401-10G
102842-310EA 383.08 USD
102842-310 102842-312
3-Chloro-4-(trifluoromethoxy)benzenesulfonyl chloride ≥95%
3-Chloro-4-(trifluoromethoxy)benzenesulfonyl chloride

Hazard: Irritant

Formula: C₇H₃Cl₂F₃O₃S
MDL Number: MFCD04972750
CAS Number: 886762-48-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR