3-Dimethylamino-2-methylpropyl chloride hydrochloride ≥97%

Supplier: Matrix Scientific

Synonyms: 3-Chloro-N,N,2-trimethylpropan-1-amine hydrochloride

084398-1G 084398-10G
102842-302EA 56.92 USD
102842-302 102842-304
3-Dimethylamino-2-methylpropyl chloride hydrochloride ≥97%
3-Dimethylamino-2-methylpropyl chloride hydrochloride

Hazard: Irritant

Formula: C₆H₁₅Cl₂N
MW: 172.093 g/mol
MDL Number: MFCD00012514
CAS Number: 4261-67-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR