1-Acetoxy-3-chloroacetone ≥97%

Supplier: Matrix Scientific

084397-1G 084397-10G
102842-298EA 156.92 USD
102842-298 102842-300
1-Acetoxy-3-chloroacetone ≥97%
1-Acetoxy-3-chloroacetone

Hazard: Irritant

Formula: C₅H₇ClO₃
MDL Number: MFCD00191365
CAS Number: 40235-68-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR