3-Bromomethylquinoxaline-2-one ≥97%

Supplier: Matrix Scientific

084391-1G 084391-10G
102842-282EA 72.31 USD
102842-282 102842-284
3-Bromomethylquinoxaline-2-one ≥97%
3-Bromomethylquinoxaline-2-one

Hazard: Irritant

Formula: C₉H₇BrN₂O
MW: 239.07 g/mol
MDL Number: MFCD03412060
CAS Number: 62235-61-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR