1-(2-Chloroethyl)-1H-hexahydroazepine hydrochloride ≥97%

Supplier: Matrix Scientific

Synonyms: 1-(2-Chloroethyl)hexamethyleneimine Hydrochloride, 1-(2-Chloroethyl)azepane hydrochloride

084387-1G 084387-10G
102842-268EA 21.54 USD
102842-268 102842-270
1-(2-Chloroethyl)-1H-hexahydroazepine hydrochloride ≥97%
1-(2-Chloroethyl)-1H-hexahydroazepine hydrochloride

Hazard: Irritant

Formula: C₈H₁₆ClN
MW: 198.13 g/mol
Melting Pt: 208…210 °C
MDL Number: MFCD00012842
CAS Number: 26487-67-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR