2-(Chloromethyl)-1,2-epoxypropane ≥97%

Supplier: Matrix Scientific

Synonyms: 2-(Chloromethyl)-2-methyloxirane, 2-(Chloromethyl)-1,2-propylene Oxide

084381-1G 084381-10G
102842-248EA 15.38 USD
102842-248 102842-250
2-(Chloromethyl)-1,2-epoxypropane ≥97%
2-(Chloromethyl)-1,2-epoxypropane

Hazard: Irritant/Store in Freezer

Formula: C₄H₇ClO
MW: 106.55 g/mol
MDL Number: MFCD00052487
CAS Number: 598-09-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR