5-(2-Bromoethyl)-2,3-dihydrobenzofuran ≥97%
Supplier: Matrix Scientific
Synonyms:
5-(2-Bromoethyl)coumaran
084376-1G
084376-5G
102842-236EA
26.15
USD
102842-236
102842-238
5-(2-Bromoethyl)-2,3-dihydrobenzofuran ≥97%
5-(2-Bromoethyl)-2,3-dihydrobenzofuran
Hazard: Irritant
Formula:
C₁₀H₁₁BrO MW: 227.10 g/mol Melting Pt: 67 °C |
MDL Number:
MFCD06797641 CAS Number: 127264-14-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...