2-Bromomethyl-1,4-benzodioxane ≥97%

Supplier: Matrix Scientific

084375-1G 084375-10G
102842-232EA 72.31 USD
102842-232 102842-234
2-Bromomethyl-1,4-benzodioxane ≥97%
2-Bromomethyl-1,4-benzodioxane

Hazard: Irritant

Formula: C₉H₉BrO₂
MW: 229.07
MDL Number: MFCD00044831
CAS Number: 2164-34-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR