5-(Bromomethyl)-benzofurazan ≥95%

Supplier: Matrix Scientific

084372-1G 084372-10G
102842-222EA 213.85 USD
102842-222 102842-224
5-(Bromomethyl)-benzofurazan ≥95%
5-(Bromomethyl)-benzofurazan

Hazard: Irritant

Formula: C₇H₅BrN₂O
MDL Number: MFCD00174369
CAS Number: 32863-31-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR