Ethyl-5-chloromethyl-2-furancarboxylate ≥97%

Supplier: Matrix Scientific

Synonyms: 5-Chloromethyl-2-furancarboxylic Acid Ethyl Ester, Ethyl 5-(chloromethyl)furan-2-carboxylate

084350-1G 084350-10G 084350-50G
102842-168EA 38.46 USD
102842-168 102842-170 102842-172
Ethyl-5-chloromethyl-2-furancarboxylate ≥97%
Ethyl-5-chloromethyl-2-furancarboxylate

Hazard: Irritant

Formula: C₈H₉ClO₃
MW: 188.61 g/mol
MDL Number: MFCD00173837
CAS Number: 2528-00-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR