1-[3-(Chloromethyl)-4-methoxyphenyl]ethanone ≥97%

Supplier: Matrix Scientific

Synonyms: 1-(3-(Chloromethyl)-4-methoxyphenyl)ethanone, 3'-(Chloromethyl)-4'-methoxyacetophenone

084325-1G 084325-10G
102842-102EA 86.15 USD
102842-102 102842-104
1-[3-(Chloromethyl)-4-methoxyphenyl]ethanone ≥97%
1-[3-(Chloromethyl)-4-methoxyphenyl]ethanone

Hazard: Irritant

Formula: C₁₀H₁₁ClO₂
MDL Number: MFCD00045267
CAS Number: 62581-82-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR