3-Methoxybenzyl bromide ≥98%

Supplier: Matrix Scientific

084317-1G 084317-50G
102842-076EA 30.77 USD
102842-076 102842-078 102842-080
3-Methoxybenzyl bromide ≥98%
3-Methoxybenzyl bromide

Hazard: Corrosive/Lachrymator

Formula: C₈H₉BrO
Boiling Pt: 152 °C
Flash Pt: 113 °C
MDL Number: MFCD00216590
CAS Number: 874-98-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR