2-Pentylphenyl isothiocyanate ≥97%

Supplier: Matrix Scientific

084316-1G 084316-10G
102842-072EA 107.69 USD
102842-072 102842-074
2-Pentylphenyl isothiocyanate ≥97%
2-Pentylphenyl isothiocyanate

Hazard: Toxic/Corrosive/Moisture Sensitive

Formula: C₁₂H₁₅NS
MW: 205.32 g/mol
Boiling Pt: 70 °C
Density: 0.93
MDL Number: MFCD00060396
CAS Number: 201224-94-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR