Benzyl-4-isocyanatotetrahydro-1(2H)-pyridinecarboxylate ≥97%

Supplier: Matrix Scientific

Synonyms: 4-Isocyanatopiperidine, N-CBZ protected

084313-1G 084313-10G
102842-058EA 410.77 USD
102842-058 102842-060
Benzyl-4-isocyanatotetrahydro-1(2H)-pyridinecarboxylate ≥97%
Benzyl-4-isocyanatotetrahydro-1(2H)-pyridinecarboxylate

Hazard: Irritant

Formula: C₁₄H₁₆N₂O₃
MW: 260.3 g/mol
MDL Number: MFCD02681988
CAS Number: 220394-91-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR