2-Thienyl isocyanate ≥85%

Supplier: Matrix Scientific

084308-1G 084308-10G
102842-046EA 170.77 USD
102842-046 102842-048
2-Thienyl isocyanate ≥85%
2-Thienyl isocyanate

Hazard: Toxic/Stored under Argon/Moisture Sensitive/Refrigerate/Lachrymator

Formula: C₅H₃NOS
Boiling Pt: 158 °C
MDL Number: MFCD02677700
CAS Number: 2048-57-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR