4-Chlorophenyl isocyanate ≥97%

Supplier: Matrix Scientific

084304-1G 084304-10G
102842-030EA 73.85 USD
102842-030 102842-032
4-Chlorophenyl isocyanate ≥97%
4-Chlorophenyl isocyanate

Hazard: Toxic/Moisture Sensitive

Formula: C₇H₄ClNO
MW: 153.57 g/mol
Boiling Pt: 203…204 °C
Melting Pt: 29…31 °C
Density: 1.2
Flash Pt: 90 °C
MDL Number: MFCD00002024
CAS Number: 104-12-1
UN: 2811
ADR: 6.1,I

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR