o-Tolyl isocyanate ≥97%

Supplier: Matrix Scientific

Synonyms: 2-Methylphenyl isocyanate, 1-Isocyanato-2-methylbenzene

084303-1G 084303-10G
102842-026EA 76.92 USD
102842-026 102842-028
o-Tolyl isocyanate ≥97%
o-Tolyl isocyanate

Hazard: Toxic/Air Sensitive/Moisture Sensitive/Lachrymator

Formula: C₈H₇NO
MW: 133.15 g/mol
Boiling Pt: 185 °C
Density: 1.074
Flash Pt: 92 °C
MDL Number: MFCD00002009
CAS Number: 614-68-6
UN: 3334
ADR: 9,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR