2-Cyclopropylethanol ≥97%
Supplier: Matrix Scientific
084281-1G
084281-10G
102841-968EA
183.08
USD
102841-968
102841-970
2-Cyclopropylethanol ≥97%
2-Cyclopropylethanol
Hazard: Flammable
Formula:
C₅H₁₀O MW: 86.13 g/mol Boiling Pt: 137-138°C Flash Pt: 47°C |
MDL Number:
MFCD00040762 CAS Number: 2566-44-1 UN: 1987 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...