3-Cyclohexene-1-methanol ≥97%

Supplier: Matrix Scientific

Synonyms: 1,2,3,6-Tetrahydrobenzyl Alcohol, Cyclohex-3-en-1-ylmethanol

084274-1G 084274-10G
102841-946EA 156.92 USD
102841-946 102841-948
3-Cyclohexene-1-methanol ≥97%
3-Cyclohexene-1-methanol

Hazard: Irritant

Formula: C₇H₁₂O
MW: 112.17 g/mol
MDL Number: MFCD00001574
CAS Number: 1679-51-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR