4-Piperidineethanol ≥97%

Supplier: Matrix Scientific

084260-1G 084260-10G
102841-916EA 127.69 USD
102841-916 102841-918
4-Piperidineethanol ≥97%
4-Piperidineethanol

Hazard: Irritant

Formula: C₇H₁₅NO
MDL Number: MFCD00006008
CAS Number: 622-26-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR