(2-Butyl-1H-imidazol-5-yl)methanol ≥97%

Supplier: Matrix Scientific

Synonyms: 2-(But-1-yl)-4-(hydroxymethyl)-1H-imidazole

084237-1G 084237-10G
102841-862EA 76.92 USD
102841-862 102841-864
(2-Butyl-1H-imidazol-5-yl)methanol ≥97%
(2-Butyl-1H-imidazol-5-yl)methanol

Hazard: Irritant

Formula: C₈H₁₄N₂O
MW: 154.21 g/mol
MDL Number: MFCD00153933
CAS Number: 68283-19-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR