(2-Butyl-5-chloro-1H-imidazol-4-yl)methanol ≥97%

Supplier: Matrix Scientific

Synonyms: 2-Butyl-5-chloro-4-hydroxymethylimidazole, (2-Butyl-4-chloro-1H-imidazol-5-yl)methanol

084236-1G 084236-10G
102841-858EA 126.15 USD
102841-858 102841-860
(2-Butyl-5-chloro-1H-imidazol-4-yl)methanol ≥97%
(2-Butyl-5-chloro-1H-imidazol-4-yl)methanol

Hazard: Irritant

Formula: C₈H₁₃ClN₂O
MW: 188.66 g/mol
MDL Number: MFCD01934397
CAS Number: 79047-41-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR