4-(Trifluoromethylthio)benzyl alcohol ≥97%

Supplier: Matrix Scientific

084201-1G 084201-10G
102841-782EA 156.92 USD
102841-782 102841-784
4-(Trifluoromethylthio)benzyl alcohol ≥97%
4-(Trifluoromethylthio)benzyl alcohol

Hazard: Irritant

Formula: C₈H₇F₃OS
MDL Number: MFCD00236342
CAS Number: 56456-52-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR