2-(3-Bromophenyl)ethanol ≥97%
Supplier: Matrix Scientific
084199-5G
084199-25G
102841-776EA
69.23
USD
102841-776
102841-778
2-(3-Bromophenyl)ethanol ≥97%
2-(3-Bromophenyl)ethanol
Hazard: Irritant-Harmful/Refrigerate
Formula:
C₈H₉BrO MW: 201.06 g/mol Boiling Pt: 107…110 °C (1 mmHg) |
MDL Number:
MFCD00191857 CAS Number: 28229-69-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...