2-Phenylethanol ≥97%

Supplier: Matrix Scientific

Synonyms: Phenethyl alcohol, Benzylcarbinol, β-Hydroxyethylbenzene, Benzyl carbinol, Benzenethanol

084198-10G 084198-50G
102841-772EA 15.38 USD
102841-772 102841-774
2-Phenylethanol ≥97%
2-Phenylethanol

Hazard: Irritant

Formula: C₆H₅CH₂CH₂OH
MW: 122.17 g/mol
Boiling Pt: 432
Flash Pt: 102
MDL Number: MFCD00002886
CAS Number: 60-12-8
UN: 2810
ADR: 6.1 III
Merck Index: 12,07370

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR