2-Phenethylbenzyl alcohol ≥97%

Supplier: Matrix Scientific

084193-1G 084193-10G
102841-760EA 156.92 USD
102841-760 102841-762
2-Phenethylbenzyl alcohol ≥97%
2-Phenethylbenzyl alcohol

Hazard: Irritant

Formula: C₁₅H₁₆O
MDL Number: MFCD00004628
CAS Number: 835-78-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR