3-Amino-2-methylbenzyl alcohol ≥97%

Supplier: Matrix Scientific

Synonyms: (3-Amino-2-methylphenyl)methanol

084190-1G 084190-10G
102841-752EA 156.92 USD
102841-752 102841-754
3-Amino-2-methylbenzyl alcohol ≥97%
3-Amino-2-methylbenzyl alcohol

Hazard: Irritant

Formula: C₈H₁₁NO
MW: 137.18 g/mol
MDL Number: MFCD00075028
CAS Number: 83647-42-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR