3-Amino-4-methylbenzyl alcohol ≥97%
Supplier: Matrix Scientific
Synonyms:
3-Amino-4-methylbenzyl Alcohol
084188-1G
084188-10G
102841-746EA
53.85
USD
102841-746
102841-748
3-Amino-4-methylbenzyl alcohol ≥97%
3-Amino-4-methylbenzyl alcohol
Hazard: Irritant
Formula:
C₈H₁₁NO MW: 137.18 g/mol |
MDL Number:
MFCD00075051 CAS Number: 81863-45-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...