5-(Hydroxymethyl)-2-iodophenol ≥97%

Supplier: Matrix Scientific

084176-1G 084176-10G
102841-720EA 241.54 USD
102841-720 102841-722
5-(Hydroxymethyl)-2-iodophenol ≥97%
5-(Hydroxymethyl)-2-iodophenol

Hazard: Irritant

Formula: C₇H₇IO₂
MDL Number: MFCD06202863
CAS Number: 773869-57-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR