2-(2-Bromoethyl)-1,3-dioxane ≥97%

Supplier: Matrix Scientific

084160-1G 084160-10G 084160-50G
102841-668EA 72.31 USD
102841-668 102841-670 102841-672
2-(2-Bromoethyl)-1,3-dioxane ≥97%
2-(2-Bromoethyl)-1,3-dioxane

Hazard: Irritant

Formula: C₆H₁₁BrO₂
MW: 195.06 g/mol
MDL Number: MFCD00006567
CAS Number: 33884-43-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR