4,7-Dichloroisatin ≥97%

Supplier: Matrix Scientific

Synonyms: 4,7-Dichloroindoline-2,3-dione, 4,7-Dichloro-1H-indole-2,3-dione

084147-1G 084147-10G
102841-630EA 61.54 USD
102841-630 102841-632
4,7-Dichloroisatin ≥97%
4,7-Dichloroisatin

Hazard: Irritant

Formula: C₈H₃Cl₂NO₂
MW: 216.02 g/mol
MDL Number: MFCD00047214
CAS Number: 18711-13-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR