5-Chloro-7-methylisatin ≥97%

Supplier: Matrix Scientific

Synonyms: 5-Chloro-7-methylindoline-2,3-dione, 5-Chloro-7-methyl-1H-indole-2,3-dione

084136-1G 084136-10G
102841-600EA 200 USD
102841-600 102841-602
5-Chloro-7-methylisatin ≥97%
5-Chloro-7-methylisatin

Hazard: Irritant

Formula: C₉H₆ClNO₂
MW: 195.6 g/mol
MDL Number: MFCD00047218
CAS Number: 14389-06-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR