2-Bromo-1-indanone ≥97%
Supplier: Matrix Scientific
084132-1G
084132-10G
102841-588EA
67.69
USD
102841-588
102841-590
2-Bromo-1-indanone ≥97%
2-Bromo-1-indanone
Hazard: Corrosive
Formula:
C₉H₇BrO Boiling Pt: 110…112 °C (0.5 hPa) Melting Pt: 33…38 °C |
MDL Number:
MFCD00041030 CAS Number: 1775-27-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...