6-Bromo-1,1,4,4-tetramethyl-1,2,3,4-tetrahydronaphthalene ≥85%

Supplier: Matrix Scientific

084099-1G 084099-10G
102841-516EA 200 USD
102841-516 102841-518
6-Bromo-1,1,4,4-tetramethyl-1,2,3,4-tetrahydronaphthalene ≥85%
6-Bromo-1,1,4,4-tetramethyl-1,2,3,4-tetrahydronaphthalene

Hazard: Irritant

Formula: C₁₄H₁₉Br
MDL Number: MFCD05664407
CAS Number: 27452-17-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR