5-Bromo-3-fluoro-2-methylpyridine ≥98%
Supplier: Matrix Scientific
083077-250MG
083077-1G
102838-858EA
369.23
USD
102838-858
102838-860
5-Bromo-3-fluoro-2-methylpyridine ≥98%
5-Bromo-3-fluoro-2-methylpyridine
Hazard: Irritant
Formula:
C₆H₅BrFN MW: 190.0100 g/mol |
MDL Number:
MFCD12196979 CAS Number: 1162674-74-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...